missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Canine GM CSF Protein

A cDNA sequence encoding the GM CSF was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP10648-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15934518

  • 4240.00 € / 1mg
Estimated Shipment: 25-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

GM CSF Protein
Research Use Only
The ED50 as calculated by the dose-dependent stimulation of the proliferation of human TF1 erythroleukemic cells is typically 1-4 ng/ml.
Canine
Untagged
GMCSF was lyophilized after extensive dialysis against 1xPBS pH 7.4.
1 mg
< 1.0 EU per ug protein as determined by the LAL method.
Recombinant Protein
E. coli
APTRSPTLVT RPSQHVDAIQ EALSLLNNSN DVTAVMNKAV KVVSEVFDPEGPTCLETRLQ LYKEGLQGSL TSLKNPLTMM ANHYKQHCPP TPESPCATQNINFKSFKENL KDFLFNIPFD CWKPVKK
Greater than 96.0% as determined by1. Analysis by RP-HPLC. 2. Analysis by SDS-PAGE.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers