missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Canine GM CSF Protein
A cDNA sequence encoding the GM CSF was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10648-2ug
Additional Details : Weight : 0.01000kg
Specifications
GM CSF Protein | |
Research Use Only | |
The ED50 as calculated by the dose-dependent stimulation of the proliferation of human TF1 erythroleukemic cells is typically 1-4 ng/ml. | |
Canine | |
Untagged | |
GMCSF was lyophilized after extensive dialysis against 1xPBS pH 7.4. |
2 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
APTRSPTLVT RPSQHVDAIQ EALSLLNNSN DVTAVMNKAV KVVSEVFDPEGPTCLETRLQ LYKEGLQGSL TSLKNPLTMM ANHYKQHCPP TPESPCATQNINFKSFKENL KDFLFNIPFD CWKPVKK | |
Greater than 96.0% as determined by1. Analysis by RP-HPLC. 2. Analysis by SDS-PAGE. |