missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Canine IFN-gamma Protein

Product Code. p-4718891
Click to view available options
Formulation:
Lyophilized from a 0.2μm filtered solution in PBS.
Lyophilized from a 0.2μm filtered solution in Sodium Succinate, Mannitol and Tween™ 80 with BSA as a carrier protein.
Unit Size:
50µg
This item is not returnable. View return policy

Product Code. 18166360

Brand: R&D Systems™ 781CG050/CF

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Canine IFN-gamma Protein has been validated for the following applications: Bioactivity.
TRUSTED_SUSTAINABILITY

Specifications

Formulation Lyophilized from a 0.2μm filtered solution in PBS.
Gene ID (Entrez) 403801
Molecular Weight (g/mol) 17 kDa
Name IFN-gamma
Quantity 50 μg
Source E. coli-derived canine IFN-gamma protein Gln24-Lys166,with an N-terminal Met
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.