missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Canine IL31 Protein
A cDNA sequence encoding the IL31 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12420-10ug
Additional Details : Weight : 0.01000kg
Specifica
100302725 | |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
Baculovirus | |
SHMAPTHQLP PSDVRKIILE LQPLSRGLLE DYQKKETGVP ESNRTLLLCL TSDSQPPRLN SSAILPYFRA IRPLSDKNII DKIIEQLDKL KFQHEPETEI SVPADTFECK SFILTILQQF SACLESVFKS LNSGPQHHHH HH | |
Greater than 95.0% as determined by SDS-PAGE. |
IL31 Protein | |
Research Use Only | |
IL-31 | |
Canine | |
His | |
IL31 Canine protein solution (1 mg/ml) containing Phosphate buffered saline (pH 7.4) and 10% glycerol. |