missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Canine LRG1 Protein
A cDNA sequence encoding the LRG1 was constructed and used to recombinantly synthesize the protein.
131.00 € - 1450.00 €
Specifications
Gene ID (Entrez) | 611876 |
---|---|
Name | LRG1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Gene Symbol | LRG1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15980889
|
enQuireBio™
QP12582-2UG |
2 μg |
131.00 €
2µg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15970889
|
enQuireBio™
QP12582-10UG |
10 μg |
203.00 €
10µg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15960889
|
enQuireBio™
QP12582-100UG |
100 μg |
1450.00 €
100µg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
611876 | |
Research Use Only | |
LRG1 | |
Canine | |
Untagged | |
LRG1 protein filtered (0.4microm) and lyophilized in 0.5 mg/ml in 20mM TRIS and 50mM NaCl, pH 8.0. |
LRG1 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
Canine | |
MSPWSPERSQ SPGGLSSHLS KTLLLLLFSV ASAQGVTPDR DTCLVFPSSN GSSVSCHPPA KLPHHFPSDT VHLVVEFFNL TQLRPDTLQG VSNLQELHLS TNQLESLSPK LLLPVPLLKV LDLTRNALTQ LPPGLFRVSA ALHTLVLKEN RLEVLEPSWL CGLKALGHLD LSGNHLQTLP PGLLANVTAL RILDLSNNQL KTLPLDLLRG PLQLERLHLE GNRLQVLEEG LLAPQPGLRY LFLSDNKLAA VAAGAFRGLQ QLDMLDLSNN SLTSVPKGLW TSLGQPTRDM EDGFDISGNP WICDHNLDDL YRWLVANKDK MFSRNDTRCA RPEALKGQTL LEAAESH | |
Purity greater than 90% as determined by SDS-PAGE. |