missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Canine TNF a Protein
A cDNA sequence encoding the TNF a was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10883-20ug
Additional Details : Weight : 0.01000kg
Specifications
403922 | |
20 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
The Specific Activity is >3.3M 105 IU/mg as determined by the cytolysis of murine L929 cells in the presence of Actinomycin D. | |
Canine | |
Untagged | |
Filtered (0.2microm) and lyophilized from a concentrated (1 mg/ml) solution in 1M PBS, pH 7.4. |
TNF a Protein | |
Research Use Only | |
TNF | |
Recombinant Protein | |
E. coli | |
VKSSSRTPSD KPVAHVVANP EAEGQLQWLS RRANALLANG VELTDNQLIV PSDGLYLIYS QVLFKGQGCP STHVLLTHTI SRFAVSYQTK VNLLSAIKSP CQRETPEGTE AKPWYEPIYL GGVFQLEKGD RLSAEINLPN YLDFAESGQV YFGIIAL | |
Greater than 95.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |