missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Cat SDF-1 beta / CXCL12 Protein
Recombinant Protein
107.00 € - 4060.00 €
Specifications
Accession Number | NP_001009847.1 |
---|---|
Name | SDF-1 beta / CXCL12 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Less than or equal to 1.00 EUs/ug as determined by the LAL method. |
Gene Symbol | CXCL12 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15985719
|
enQuireBio™
QP5218-2UG |
2 μg |
107.00 €
2µg |
Estimated Shipment: 28-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15965719
|
enQuireBio™
QP5218-10UG |
10 μg |
252.00 €
10µg |
Estimated Shipment: 28-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15955719
|
enQuireBio™
QP5218-100UG |
100 μg |
630.00 €
100µg |
Estimated Shipment: 28-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15995719
|
enQuireBio™
QP5218-500UG |
500 μg |
2490.00 €
500µg |
Estimated Shipment: 28-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15975719
|
enQuireBio™
QP5218-1MG |
1 mg |
4060.00 €
1mg |
Estimated Shipment: 28-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
NP_001009847.1 | |
Research Use Only | |
CXCL12 | |
Cat | |
Untagged | |
10 mM sodium phosphate, pH 7.5 |
SDF-1 beta / CXCL12 Protein | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
KPVSLSYRCP CRFFESHVAR ANVKHLKILN TPNCALQIVA RLKNNNRQVC IDPKLKWIQE YLEKALNKRF KM | |
Greater than 95% as determined by SDS-PAGE. |