missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Chicken Focal adhesion kinase 1 Protein
A DNA sequence encoding the Gallus gallus (Chicken) Focal adhesion kinase 1, was expressed in the hosts and tags indicated.
309.00 € - 3260.00 €
Specifications
Accession Number | NP_990766.1 |
---|---|
Gene ID (Entrez) | 396416 |
Name | Focal adhesion kinase 1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16046531
|
enQuireBio™
QP8800-EC-100UG |
100 μg |
940.00 €
100µg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16066531
|
enQuireBio™
QP8800-EC-1MG |
1 mg |
3260.00 €
1mg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16056531
|
enQuireBio™
QP8800-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16076531
|
enQuireBio™
QP8800-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16086531
|
enQuireBio™
QP8800-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16096531
|
enQuireBio™
QP8800-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_990766.1 | |
Focal adhesion kinase 1 Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Chicken | |
GST | |
Tris-based buffer, 50% glycerol |
396416 | |
Research Use Only | |
PTK2 | |
Recombinant Protein | |
E. coli | |
LANNEKQGVR SHTVSVSETD DYAEIIDEED TYTMPSTRDY EIQRERIELG RCIGEGQFGD VHQGIYMSPE NPAMAVAIKT CKNCTSDSVR EKFLQEALTM RQFDHPHIVK LIGVITENPV WIIMELCTLG ELRSFLQVRK FSLDLASLIL YAYQLSTALA YLESKRFVHR DIAARNVLVS ATDCVKLGDF GLSRYMEDST YYKASKGKLP IKWMAPESIN FRRFTSASDV WMFGVCMWEI LMHGVKPFQG VKNNDVIGRI ENGERLPMPP NCPPTLYSLM TKCWAYDPSR RPRFTELKAQ LSTILEEE | |
Greater than 90% as determined by SDS-PAGE. |