missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Chicken Leptin Protein

A cDNA sequence encoding the Leptin was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP10747-20ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15955048

  • 131.00 € / 20µg
Estimated Shipment: 24-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

Leptin Protein
Research Use Only
Biological active as evidenced by inducing proliferation of BAF/3 cells stably transfected with the long form of human leptin receptor. Its activity is however 5-10 fold lower as compared to mammalian leptins.
Chicken
Untagged
The protein was lyophilized from a concentrated (1 mg/ml) solution with 0.0045mM NaHCO3.
20 μg
< 1.0 EU per ug protein as determined by the LAL method.
Recombinant Protein
E. coli
MCWRPLCRLW SYLVYVQAVP CQIFQDDTKT LIKTIVTRIN DISHTSVSAK QRVTGLDFIP GLHPILSLSK MDQTLAVYQQ VLTSLPSQNV LQIANDLENL RDLLHLLAFS KSCSLPQTSG LQKPESLDGV LEASLYSTEV VALSRLQGSL QDILQQLDIS PEC
Greater than 95.0% as determined by:(a) Analysis by SEC-HPLC. (b) Analysis by SDS-PAGE.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers