missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Chicken Vitellogenin-1 Protein

A DNA sequence encoding the Gallus gallus (Chicken) Vitellogenin-1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6991-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16046940

  • 2260.00 € / 500µg
Estimated Shipment: 31-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001004408.1
Vitellogenin-1 Protein
Research Use Only
VTG1
Recombinant Protein
E. coli
FGENKVYTYN YESILFSGIP EKGLARTGIR IRSEVEISGI GPKLCLIRIH SIEAAEYNGI WPTSSFSRSL KLTQALTGQL SIPIKFEYSN GHVGNLMAPD SVSDDGLNIY RGILNILELS LKKMQHSYSI QEAGIGGICN TTYAIQENKK ANLVDVTKSK DLNSCEEKVQ VVTGSAYTQP CQTCQQRNKN SRATATYNYK IKYTHNEAVI TQAEVEEVHQ FTPFHEITGG NAIVEARQKL ALIEVQKQVA EVPPKEFQKR GSLQYQFGSE LLQLPVHLFK IKDVERQIEE RLQDLVETTY EQLPSDAPAK ALKLMHLLRA ANEENYESVW KQFSSRPAYR RYLLDLLPAA ASHRSLRFLR HKMERQELTN WEIAQTVLVA LHSSSPTQEV MEEATLIVKK HCPRSSSVLR KVCLLSYASL CHKRCSSPYS CSECLQVFHV FAGEALGKSN IEEVLLALKA LGNVGHPASI KHIKKFLPGY AAGASELPLK VHETAVMALK SIGMRDPQMV QAITLEIFLN HKIHPRIRML AAVVLLETKP GLPILMILVD AVLKEPSMQV ASFIYSHLRA LGRSTAPDLQ MMASACRMAV RALSPKFDRS GYQFSKVFRF
Greater than 90% as determined by SDS-PAGE.
424547
500 μg
Not determined.
Full Length
Chicken
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers