missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Chlamydia PGP-3D Protein

A cDNA sequence encoding the Chlamydia PGP-3D was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11412-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15907188

  • 228.00 € / 100µg
Estimated Shipment: 25-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

PGP-3D Protein
Research Use Only
Recombinant Protein
E. coli
10mM Tris-HCl, pH 8.0, 1.5M Urea and 50% Glycerol.
100 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Chlamydia
His
Protein is >90% pure as determined by SDS-PAGE.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers