missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Chlamydia PGP-3D Protein
A cDNA sequence encoding the Chlamydia PGP-3D was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11412-500ug
Additional Details : Weight : 0.01000kg
Specifications
PGP-3D Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
10mM Tris-HCl, pH 8.0, 1.5M Urea and 50% Glycerol. |
500 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Chlamydia | |
His | |
Protein is >90% pure as determined by SDS-PAGE. |