missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Chlamydia W2 Protein
A cDNA sequence encoding the Chlamydia W2 was constructed and used to recombinantly synthesize the protein.
228.00 € - 1740.00 €
Specifications
Name | W2 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Chlamydia |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15967188
|
enQuireBio™
QP11414-100UG |
100 μg |
228.00 €
100µg |
Estimated Shipment: 27-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15987188
|
enQuireBio™
QP11414-500UG |
500 μg |
870.00 €
500µg |
Estimated Shipment: 27-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15977188
|
enQuireBio™
QP11414-1MG |
1 mg |
1740.00 €
1mg |
Estimated Shipment: 27-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
W2 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Chlamydia | |
Untagged | |
Protein is >90% pure as determined by SDS-PAGE. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
20mM Tris-HCl, pH 7.2, 50% glycerol and 1.5M urea. |