missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Cynomolgus CD155/PVR Fc Chimera Protein
Description
Measured by its binding ability in a functional ELISA. When Recombinant Cynomolgus Monkey CD155/PVR Fc Chimera is coated at 5 μg/mL (100 μL/well), the concentration of Recombinant Human CD96v2 Fc Chimera (Catalog # 9556-CD) that produces 50% optimal binding response is 0.25-1.25 μg/mL.
Specifications
Specifications
| Accession Number | unique sequence |
| For Use With (Application) | Bioactivity |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. |
| Gene ID (Entrez) | 5817 |
| Molecular Weight (g/mol) | M.W. (Observed): 68-77 kDa, reducing conditions; M.W. (theoretical): 85 kDa |
| Purification Method | >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining |
| Quantity | 50 μg |
| Source | Human embryonic kidney cell, HEK293-derived cynomolgus monkey CD155/PVR protein Cynomolgus Monkey CD155/PVR (Asp28-Asn343) (N-terminus) Accession # XP_011734201.1 (T77M, V93T, G223S, R328G) IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, <= -20 °C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <0.10 EU / 1 μg of the protein by the LAL method. |
| Show More |
Nom du produit
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.
Vous avez repéré une opportunité d'amélioration ?