missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Cynomolgus LILRB5/CD85c/LIR-8 Fc Protein
Description
Measured by its binding ability in a functional ELISA. When Recombinant Cynomolgus Monkey LILRB5/CD85c/LIR-8 Fc Chimera (Catalog # 11102-T4) is immobilized at 2 μg/mL (100 μL/well), Recombinant Human Angiopoietin-like Protein 7/ANGPTL7 (Catalog # 914-AN ) binds with an ED50 of 14.0-140 ng/mL.
Specifications
Specifications
| Accession Number | XP_005590354.1 |
| For Use With (Application) | Bioactivity |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. |
| Gene ID (Entrez) | 10990 |
| Molecular Weight (g/mol) | M.W.-observed: 86-99 kDa, under reducing conditions., M.W.-theroretical: 74 kDa |
| Quantity | 50 μg |
| Source | Chinese Hamster Ovary cell line, CHO-derived cynomolgus monkey LILRB5/CD85c/LIR-8 protein Cynomolgus Monkey LILRB5/CD85c/LIR-8 (Gly24-Gly458) (C-terminus) Accession # XP_005590354.1 IEGRMD Human IgG1 (Pro100-Lys330) (N-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 week, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -70°C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <0.10 EU / 1 μg of the protein by the LAL method. |
| Gene Alias | CD85 antigen-like family member C, CD85c, CD85c antigen, Leukocyte immunoglobulin-like receptor 8, leukocyte immunoglobulin-like receptor, subfamily B (with TM and ITIM domains), LIR8, LIR-8, LIR8CD85C, LIR-8 leukocyte immunoglobulin-like receptor subfamily B member 5, member 5 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?