missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Cynomolgus Monkey Klotho beta His Protein
Click to view available options
Quantity:
25 μg
Unit Size:
25µg
Description
Measured by its binding ability in a functional ELISA. When Recombinant Human FGF-21 (Catalog # 2539-FG ) is immobilized at 1 μg/mL (100 μL/well), Recombinant Cynomolgus Monkey Klotho beta His-tag (Catalog # 10428-KB) binds with an ED50 of 0.05-0.4 μg/mL.
Specifications
Specifications
| Accession Number | EHH53620.1 |
| For Use With (Application) | Bioactivity |
| Formulation | Supplied as a 0.2 μm filtered solution in PBS, Glycerol and EDTA. |
| Gene ID (Entrez) | 152831 |
| Molecular Weight (g/mol) | M.W. (Observed): 110-130 kDa, under reducing conditions; M.W. (theoretical): 110 kDa |
| Quantity | 25 μg |
| Source | Mouse myeloma cell line, NS0-derived cynomolgus monkey Klotho beta protein Phe53-Leu997, with a C-terminal 6-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -70 °C as supplied. 3 months, -70 °C under sterile conditions after opening. 1 month, 2°C to 8°C under sterile conditions after opening. |
| Endotoxin Concentration | <0.10 EU / 1 μg of the protein by the LAL method. |
| Conjugate | Unconjugated |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction