missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Cynomolgus/Rhesus Macaque BTLA Fc Protein
Description
Measured by its binding ability in a functional ELISA. When Recombinant Mouse HVEM/TNFRSF14 Fc Chimera (Catalog # 2516-HV ) is immobilized at 0.5 μg/mL, Recombinant Cynomolgus Monkey BTLA Fc Chimera binds with an ED50 of 25-125 ng/mL.
Specifications
Specifications
| Accession Number | XP_005548224 |
| For Use With (Application) | Bioactivity |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. |
| Gene ID (Entrez) | 151888 |
| Molecular Weight (g/mol) | M.W. (Observed): 48-58 kDa, reducing conditions; M.W. (theoretical): 40.3 kDa |
| Quantity | 50 μg |
| Source | Human embryonic kidney cell, HEK293-derived BTLA protein Cynomolgus Monkey BTLA (Met1-Leu150) (N-terminus) Accession # XP_005548224 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus) |
| Storage Requirements | 12 months from date of receipt, <= -20 °C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, <= -20 °C under sterile conditions after reconstitution. |
| Regulatory Status | RUO |
| Endotoxin Concentration | <0.10 EU / 1 μg of the protein by the LAL method. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?