missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Dengue Virus Dengue Envelope-4 Protein
A cDNA sequence encoding the Dengue Envelope-4 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11635-1mg
Specifications
Dengue Envelope-4 Protein | |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
Dengue Envelope-4 protein solution in 1xD-PBS, pH 7.4, 0.1% Thimerosal, 5mM EDTA, 1microg/ml of Leupeptin, Aprotinin and Pepstatin A. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Dengue Virus | |
Untagged | |
Protein is >95% pure as determined by 12.5% SDS-PAGE. |