missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Dengue Virus Dengue p18 Protein

A cDNA sequence encoding the Dengue p18 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11648-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15967908

  • 1740.00 € / 1mg
Estimated Shipment: 07-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

Dengue p18 Protein
Research Use Only
Recombinant Protein
E. coli
Phosphate buffered saline, pH 7.4 and 0.02% sodium nitrate.
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Dengue Virus
Untagged
Protein is >95% pure as determined by 12% PAGE (coomassie staining).
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions