missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Dengue Virus Serotype 2 VLP Protein
Recombinant Protein
1025.00 € - 4470.00 €
Specifications
Name | Serotype 2 VLP Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Untested |
Product Type | Recombinant Protein |
Cross Reactivity | Dengue virus (Strain: Thailand/16681/1984) |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15933998
|
enQuireBio™
QP10479-100UG |
100 μg |
1025.00 €
100µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15943998
|
enQuireBio™
QP10479-500UG |
500 μg |
4470.00 €
500µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This VLP protein is combinantion of the C, E1 and E2 sequences. Virus-Like Particles (VLPs) are a new form of protein expression that consist of protein shells with outer proteins that are specific to the given virus. VLPs are immunogenic but are non-infectious, lacking the primary genetic material of the virus. Additionally, VLPs more effectively activate aspects of the immune response, which improves immune stimulation and immunological memory. VLP-based vaccines have also shown to provide effective protection and are in current use for several diseases.Specifications
Serotype 2 VLP Protein | |
Untested | |
Dengue virus (Strain: Thailand/16681/1984) | |
Native | |
>95% as determined by SDS-PAGE. |
Research Use Only | |
Recombinant Protein | |
Human | |
10mM sodium phosphate, 20mM sodium citrate, 154mM sodium chloride, pH7.4. |