missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Denis GH Protein
A cDNA sequence encoding the GH was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11961-10ug
Additional Details : Weight : 0.01000kg
Specifications
GH Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MDRVVLMLSV MSLGVSSQPI TDGQRLFSIA VSRVQHLHLL AQRLFSDFES SLQTEEQRQL NKIFLQDFCN SDYIISPIDK HETQRSSVLK LLSISYRLVE SWEFPSRSLS GGSAPRNQIS PKLSELKTGI HLLIRANEDG AEIFPDSSAL QLAPYGNYYQ SLGTDESLRR TYELLACFKK DMHKVETYLT VAKCRLSPEA NCTL | |
Greater than 97.0% as determined by:(a) Analysis by SEC-HPLC. (b) Analysis by SDS-PAGE. |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Denis | |
Untagged | |
The protein was lyophilized from a concentrated (1 mg/ml) solution with 0.0045mM NaHCO3. |