missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Dog CALCA / CGRP Protein
A DNA sequence encoding the Canis lupus familiaris (Dog) (Canis familiaris) CALCA / CGRP, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5746-ec-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001003266.1 | |
CALCA / CGRP Protein | |
Research Use Only | |
CALCA | |
Recombinant Protein | |
E. coli | |
CSNLSTCVLG TYSKDLNNFH TFSGIGFGAE TP | |
Greater than 90% as determined by SDS-PAGE. |
403946 | |
100 μg | |
Not determined. | |
Full Length | |
Dog | |
His-SUMO | |
Tris-based buffer, 50% glycerol |