missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Dog Cationic trypsin Protein

Product Code. p-7155887
Change view
Click to view available options
Quantity:
10 μg
100 μg
50 μg
Unit Size:
100µg
10µg
50µg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16029161 10 μg 10µg
16039161 50 μg 50µg
16019161 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16029161 Supplier enQuireBio™ Supplier No. QP9998iv10ug

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

A DNA sequence encoding the Canis lupus familiaris (Dog) (Canis familiaris) Cationic trypsin, was expressed in the hosts and tags indicated.

Specifications

Name Cationic trypsin Protein
Quantity 10 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Protein Length Full Length
Product Type Recombinant Protein
Cross Reactivity Dog
Species E. coli
Protein Tag His-SUMO
Sequence IVGGYTCSRN SVPYQVSLNS GYHFCGGSLI NSQWVVSAAH CYKSRIQVRL GEYNIAVSEG GEQFINAAKI IRHPRYNANT IDNDIMLIKL SSPATLNSRV SAIALPKSCP AAGTQCLISG WGNTQSIGQN YPDVLQCLKA PILSDSVCRN AYPGQISSNM MCLGYMEGGK DSCQGDSGGP VVCNGELQGV VSWGAGCAQK GKPGVSPKVC KYVSWIQQTI AAN
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 85% as determined by SDS-PAGE.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.