missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Dog CMA1 / Chymase 1 Protein
A DNA sequence encoding the Canis lupus familiaris (Dog) (Canis familiaris) CMA1 / Chymase 1, was expressed in the hosts and tags indicated.
309.00 € - 3260.00 €
Specifications
Accession Number | NP_001013442.1 |
---|---|
Gene ID (Entrez) | 490628 |
Name | CMA1 / Chymase 1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15908179
|
enQuireBio™
QP5843-EC-100UG |
100 μg |
940.00 €
100µg |
Estimated Shipment: 27-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15928179
|
enQuireBio™
QP5843-EC-1MG |
1 mg |
3260.00 €
1mg |
Estimated Shipment: 27-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15918179
|
enQuireBio™
QP5843-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15938179
|
enQuireBio™
QP5843-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15948179
|
enQuireBio™
QP5843-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15958179
|
enQuireBio™
QP5843-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001013442.1 | |
CMA1 / Chymase 1 Protein | |
Not determined. | |
Full Length | |
Dog | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
490628 | |
Research Use Only | |
CMA1 | |
Recombinant Protein | |
E. coli | |
IIGGTESKPH SRPYMAHLEI LTLRNHLASC GGFLIRRNFV LTAAHCAGRF IMVTLGAHNI QKKEDTWQKL EVIKQFPHPK YDDLTLRHDI MLLKLKEKAN LTLAVGTLPL SPQFNFVPPG RMCRVAGWGK RQVNGSGSDT LQEVKLRLMD PQACRHYMAF DHNLQLCVGN PRKTKSAFKG DSGGPLLCAG VAQGIVSYGQ NDAKPPAVFT RISHYRPWIN KVLKQNKA | |
Greater than 90% as determined by SDS-PAGE. |