missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Dog DMP1 Protein
A DNA sequence encoding the Canis lupus familiaris (Dog) (Canis familiaris) DMP1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6934-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
DMP1 Protein | |
Research Use Only | |
DMP1 | |
Recombinant Protein | |
E. coli | |
MHRPAGGLSR SGGKEGDDKD DDEDDSGDDT FGDDDNGPGP EERQGGKSRL KSNEDSADST QSREDSPSLG EDSAQDTTSE SRDLDNEDEM NSSPERGDSV QESESKEHWV GGGSEGDSSH GDGSEFGDEE MQNDDPDTIR NERGNSRMNS ASVKSEESKG NSDEQASTQD SGDSQSMEYP RRKIFRKSRI SEEDDIGELD DSNTMEEVKS DSTENANSKE TGLSQSRENS ESESVEESEE NQSQEDSQNI QDPSSESSQE VDLPSQENSS ESQEEVVTES RGDNPDNTTS HSDQEDSDSS EENSLNKPSS SESESTEEQA DSESNESLRS SEESPESTEE NSSSQEDLQS HSASAESQSE ESQSEQDSQS EEDDASDSQD SSKLKEDSNS TESKSSSEEE GQSKNTEIES RKLIVDTYHN KPIGDQDDND CQDGY | |
Greater than 90% as determined by SDS-PAGE. |
1 mg | |
Not determined. | |
Full Length | |
Dog | |
His-SUMO | |
Tris-based buffer, 50% glycerol |