missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Dog IL-8 / CXCL8 Protein

A DNA sequence encoding the Canis familiaris (Dog) (Canis lupus familiaris) IL-8 / CXCL8, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6227-ye-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16031760

  • 3900.00 € / 1mg
Estimated Shipment: 24-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001003200.1
IL-8 / CXCL8 Protein
Research Use Only
CXCL8
Recombinant Protein
Yeast
AVLSRVSSEL RCQCIKTHST PFHPKYIKEL RVIDSGPHCE NSEIIVKLFN GNEVCLDPKE KWVQKVVQIF LKKAEKQDP
Greater than 90% as determined by SDS-PAGE.
403850
1 mg
Not determined.
Full Length
Dog
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers