missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli 50S ribosomal protein L30 Protein
Specifications
Specifications
| Accession Number | NP_417761.1 |
| Gene ID (Entrez) | 947797 |
| Name | 50S ribosomal protein L30 Protein |
| Quantity | 500 μg |
| Regulatory Status | Research Use Only |
| Endotoxin Concentration | Not determined. |
| Gene Symbol | rpmD |
| Protein Length | Full Length |
| Product Type | Recombinant Protein |
| Cross Reactivity | E. coli |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?