missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant E. coli 50S ribosomal protein L9 Protein

A DNA sequence encoding the Escherichia coli (strain K12) 50S ribosomal protein L9, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8623-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16035631

  • 967.00 € / 100µg
Estimated Shipment: 03-08-2022
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_418624.1
50S ribosomal protein L9 Protein
100 μg
Not determined.
Partial (see sequence information for more details).
E. coli
GST
Tris-based buffer, 50% glycerol
948720
Greater than 90% as determined by SDS-PAGE.
Research Use Only
rplI
Recombinant Protein
E. coli
MQVILLDKVA NLGSLGDQVN VKAGYARNFL VPQGKAVPAT KKNIEFFEAR RAELEAKLAE VLAAANARAE KINALETVTI ASKAGDEGKL FGSIGTRDIA DAVTAAGVEV AKSEVRLPNG VLRTTGEHEV SFQVHSEVFA KVIV
Videos
SDS
Documents
Certificates
Special Offers

Special Offers