missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli Cell division protein ZipA Protein
A DNA sequence encoding the Escherichia coli (strain K12) Cell division protein ZipA, was expressed in the hosts and tags indicated.
421.00 € - 3590.00 €
Specifications
Accession Number | NP_416907.1 |
---|---|
Gene ID (Entrez) | 946869 |
Name | Cell division protein ZipA Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16036890
|
enQuireBio™
QP6957-10UG |
10 μg |
421.00 €
10µg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16076890
|
enQuireBio™
QP6957-50UG |
50 μg |
605.00 €
50µg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16026890
|
enQuireBio™
QP6957-100UG |
100 μg |
1025.00 €
100µg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16056890
|
enQuireBio™
QP6957-200UG |
200 μg |
1690.00 €
200µg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16066890
|
enQuireBio™
QP6957-500UG |
500 μg |
2260.00 €
500µg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16046890
|
enQuireBio™
QP6957-1MG |
1 mg |
3590.00 €
1mg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
NP_416907.1 | |
Cell division protein ZipA Protein | |
Not determined. | |
Full Length | |
E. coli | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
946869 | |
Research Use Only | |
zipA | |
Recombinant Protein | |
E. coli | |
MMQDLRLILI IVGAIAIIAL LVHGFWTSRK ERSSMFRDRP LKRMKSKRDD DSYDEDVEDD EGVGEVRVHR VNHAPANAQE HEAARPSPQH QYQPPYASAQ PRQPVQQPPE AQVPPQHAPH PAQPVQQPAY QPQPEQPLQQ PVSPQVAPAP QPVHSAPQPA QQAFQPAEPV AAPQPEPVAE PAPVMDKPKR KEAVIIMNVA AHHGSELNGE LLLNSIQQAG FIFGDMNIYH RHLSPDGSGP ALFSLANMVK PGTFDPEMKD FTTPGVTIFM QVPSYGDELQ NFKLMLQSAQ HIADEVGGVV LDDQRRMMTP QKLREYQDII REVKDANA | |
Greater than 90% as determined by SDS-PAGE. |