missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant E. coli CFA / I fimbrial subunit E Protein

A DNA sequence encoding the Escherichia coli CFA/I fimbrial subunit E, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7131-ye-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16047470

  • 1050.00 € / 100µg
Estimated Shipment: 27-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

WP_001033658.1
100 μg
Not determined.
Full Length
E. coli
His
Tris-based buffer, 50% glycerol
CFA / I fimbrial subunit E Protein
Research Use Only
cfaE
Recombinant Protein
Yeast
MNKILFIFTL FFSSGFFTFA VSADKNPGSE NMTNTIGPHD RGGSSPIYNI LNSYLTAYNG SHHLYDRMSF LCLSSQNTLN GACPSSDAPG TATIDGETNI TLQFTEKRSL IKRELQIKGY KQFLFKNANC PSKLALNSSH FQCNREQASG ATLSLYIPAG ELNKLPFGGV WNAVLKLNVK RRYDTTYGTY TINITVNLTD KGNIQIWLPQ FKSNARVDLN LRPTGGGTYI GRNSVDMCFY DGYSTNSSSL EIRFQDDNSK SDGKFYLKKI NDDSKELVYT LSLLLAGKNL TPTNGQALNI NTASLETNWN RITAVTMPEI SVPVLCWPGR LQLDAKVKNP EAGQYMGNIK ITFTPSSQTL
Greater than 80% as determined by SDS-PAGE.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers