missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant E. coli CS6 fimbrial subunit B Protein

Product Code. 16067820
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16067820 10 μg 10µg
16007830 50 μg 50µg
16057820 100 μg 100µg
16087820 200 μg 200µg
16097820 500 μg 500µg
16077820 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 16067820 Supplier enQuireBio™ Supplier No. QP7239ec10ug

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

A DNA sequence encoding the Escherichia coli (strain K12) CS6 fimbrial subunit B, was expressed in the hosts and tags indicated.

Specifications

Accession Number WP_000913743.1
Name CS6 fimbrial subunit B Protein
Quantity 10 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol cssB
Protein Length Full Length
Product Type Recombinant Protein
Cross Reactivity E. coli
Species E. coli
Protein Tag His-SUMO
Sequence GNWQYKSLDV NVNIEQNFIP DIDSAVRIIP VNYDSDPKLN SQLYTVEMTI PAGVSAVKIV PTDSLTSSGQ QIGKLVNVNN PDQNMNYYIR KDSGAGKFMA GQKGSFSVKE NTSYTFSAIY TGGEYPNSGY SSGTYAGHLT VSFYSN
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 92.2% as determined by SDS-PAGE.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.