missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli Cytosine deaminase Protein
A DNA sequence encoding the Escherichia coli (strain K12) Cytosine deaminase, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7203-ye-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_414871.1 | |
Cytosine deaminase Protein | |
Research Use Only | |
codA | |
Recombinant Protein | |
Yeast | |
SNNALQTIIN ARLPGEEGLW QIHLQDGKIS AIDAQSGVMP ITENSLDAEQ GLVIPPFVEP HIHLDTTQTA GQPNWNQSGT LFEGIERWAE RKALLTHDDV KQRAWQTLKW QIANGIQHVR THVDVSDATL TALKAMLEVK QEVAPWIDLQ IVAFPQEGIL SYPNGEALLE EALRLGADVV GAIPHFEFTR EYGVESLHKT FALAQKYDRL IDVHCDEIDD EQSRFVETVA ALAHHEGMGA RVTASHTTAM HSYNGAYTSR LFRLLKMSGI NFVANPLVNI HLQGRFDTYP KRRGITRVKE MLESGINVCF GHDDVFDPWY PLGTANMLQV LHMGLHVCQL MGYGQINDGL NLITHHSART LNLQDYGIAA GNSANLIILP AENGFDALRR QVPVRYSVRG GKVIASTQPA QTTVYLEQPE AIDYKR | |
Greater than 90% as determined by SDS-PAGE. |
944996 | |
100 μg | |
Not determined. | |
Full Length | |
E. coli | |
His | |
Tris-based buffer, 50% glycerol |