missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant E. coli DNA ligase Protein

A DNA sequence encoding the Escherichia coli (strain K12) DNA ligase, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7075-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16037290

  • 1025.00 € / 100µg
Estimated Shipment: 06-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_416906.1
DNA ligase Protein
Research Use Only
ligA
Recombinant Protein
E. coli
MESIEQQLTE LRTTLRHHEY LYHVMDAPEI PDAEYDRLMR ELRELETKHP ELITPDSPTQ RVGAAPLAAF SQIRHEVPML SLDNVFDEES FLAFNKRVQD RLKNNEKVTW CCELKLDGLA VSILYENGVL VSAATRGDGT TGEDITSNVR TIRAIPLKLH GENIPARLEV RGEVFLPQAG FEKINEDARR TGGKVFANPR NAAAGSLRQL DPRITAKRPL TFFCYGVGVL EGGELPDTHL GRLLQFKKWG LPVSDRVTLC ESAEEVLAFY HKVEEDRPTL GFDIDGVVIK VNSLAQQEQL GFVARAPRWA VAFKFPAQEQ MTFVRDVEFQ VGRTGAITPV ARLEPVHVAG VLVSNATLHN ADEIERLGLR IGDKVVIRRA GDVIPQVVNV VLSERPEDTR EVVFPTHCPV CGSDVERVEG EAVARCTGGL ICGAQRKESL KHFVSRRAMD VDGMGDKIID QLVEKEYVHT PADLFKLTAG KLTGLERMGP KSAQNVVNAL EKAKETTFAR FLYALGIREV GEATAAGLAA YFGTLEALEA ASIEELQKVP DVGIVVASHV HNFFAEESNR NVISELLAEG VHWPAPIVIN AEEIDSPFAG KTVVLTGSLS QMSRDDAKAR LVELGAKVAG SVSKKTDLVI AGEAAGSKLA KAQELGIEVI DEAEMLRLLG S
Greater than 90% as determined by SDS-PAGE.
946885
100 μg
Not determined.
Full Length
E. coli
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers