missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli Glutamate--cysteine ligase Protein
A DNA sequence encoding the Escherichia coli (strain K12) Glutamate--cysteine ligase, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7387-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_417173.1 | |
Glutamate--cysteine ligase Protein | |
1 mg | |
Not determined. | |
Full Length | |
E. coli | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
944881 | |
Greater than 91.2% as determined by SDS-PAGE. | |
Research Use Only | |
gshA | |
Recombinant Protein | |
E. coli | |
MIPDVSQALA WLEKHPQALK GIQRGLERET LRVNADGTLA TTGHPEALGS ALTHKWITTD FAEALLEFIT PVDGDIEHML TFMRDLHRYT ARNMGDERMW PLSMPCYIAE GQDIELAQYG TSNTGRFKTL YREGLKNRYG ALMQTISGVH YNFSLPMAFW QAKCGDISGA DAKEKISAGY FRVIRNYYRF GWVIPYLFGA SPAICSSFLQ GKPTSLPFEK TECGMYYLPY ATSLRLSDLG YTNKSQSNLG ITFNDLYEYV AGLKQAIKTP SEEYAKIGIE KDGKRLQINS NVLQIENELY APIRPKRVTR SGESPSDALL RGGIEYIEVR SLDINPFSPI GVDEQQVRFL DLFMVWCALA DAPEMSSSEL ACTRVNWNRV ILEGRKPGLT LGIGCETAQF PLPQVGKDLF RDLKRVAQTL DSINGGEAYQ KVCDELVACF DNPDLTFSAR ILRSMIDTGI GGTGKAFAEA YRNLLREEPL EILREEDFVA EREASERRQQ EMEAADTEPF AVWLEKHA |