missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant E. coli Glutaminase 1 Protein

A DNA sequence encoding the Escherichia coli (strain K12) Glutaminase 1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6951-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16056850

  • 288.00 € / 50µg
Estimated Shipment: 27-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_415018.1
Glutaminase 1 Protein
Research Use Only
glsA1
Recombinant Protein
E. coli
MLDANKLQQA VDQAYTQFHS LNGGQNADYI PFLANVPGQL AAVAIVTCDG NVYSAGDSDY RFALESISKV CTLALALEDV GPQAVQDKIG ADPTGLPFNS VIALELHGGK PLSPLVNAGA IATTSLINAE NVEQRWQRIL HIQQQLAGEQ VALSDEVNQS EQTTNFHNRA IAWLLYSAGY LYCDAMEACD VYTRQCSTLL NTIELATLGA TLAAGGVNPL THKRVLQADN VPYILAEMMM EGLYGRSGDW AYRVGLPGKS GVGGGILAVV PGVMGIAAFS PPLDEDGNSV RGQKMVASVA KQLGYNVFKG
Greater than 90% as determined by SDS-PAGE.
946187
50 μg
Not determined.
Full Length
E. coli
His-GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers