missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli Long-chain fatty acid transport Protein
A DNA sequence encoding the Escherichia coli O157:H7 Long-chain fatty acid transport protein, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7835-ec-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_311254.2 | |
Long-chain fatty acid transport Protein | |
Research Use Only | |
fadL | |
Recombinant Protein | |
E. coli | |
AGFQLNEFSS SGLGRAYSGE GAIADDAGNV SRNPALITMF DRPTFSAGAV YIDPDVNISG TSPSGRSLKA DNIAPTAWVP NMHFVAPIND QFGWGASITS NYGLATEFND TYAGGSVGGT TDLETMNLNL SGAYRLNNAW SFGLGFNAVY ARAKIERFAG DLGQLVAGQI MQSPAGKTPQ GQALAATANG IDSNTKIAHL NGNQWGFGWN AGILYELDKN NRYALTYRSE VKIDFKGNYS SDLNRVFNNY GLPIPTATGG ATQSGYLTLN LPEMWEVSGY NRVDPQWAIH YSLAYTSWSQ FQQLKATSTS GDTLFQKHEG FKDAYRIALG TTYYYDDNWT FRTGIAFDDS PVPAQNRSIS IPDQDRFWLS AGTTYAFNKD ASVDVGVSYM HGQSVKINEG PYQFESEGKA WLFGTNFNYA F | |
Greater than 90% as determined by SDS-PAGE. |
915676 | |
100 μg | |
Not determined. | |
Full Length | |
E. coli | |
His-SUMO | |
Tris-based buffer, 50% glycerol |