missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli Malate synthase G Protein
A DNA sequence encoding the Escherichia coli (strain K12) Malate synthase G, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7126-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_417450.1 | |
Malate synthase G Protein | |
1 mg | |
Not determined. | |
Full Length | |
E. coli | |
His | |
Tris-based buffer, 50% glycerol |
948857 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
glcB | |
Recombinant Protein | |
E. coli | |
SQTITQSRLR IDANFKRFVD EEVLPGTGLD AAAFWRNFDE IVHDLAPENR QLLAERDRIQ AALDEWHRSN PGPVKDKAAY KSFLRELGYL VPQPERVTVE TTGIDSEITS QAGPQLVVPA MNARYALNAA NARWGSLYDA LYGSDIIPQE GAMVSGYDPQ RGEQVIAWVR RFLDESLPLE NGSYQDVVAF KVVDKQLRIQ LKNGKETTLR TPAQFVGYRG DAAAPTCILL KNNGLHIELQ IDANGRIGKD DPAHINDVIV EAAISTILDC EDSVAAVDAE DKILLYRNLL GLMQGTLQEK MEKNGRQIVR KLNDDRHYTA ADGSEISLHG RSLLFIRNVG HLMTIPVIWD SEGNEIPEGI LDGVMTGAIA LYDLKVQKNS RTGSVYIVKP KMHGPQEVAF ANKLFTRIET MLGMAPNTLK MGIMDEERRT SLNLRSCIAQ ARNRVAFINT GFLDRTGDEM HSVMEAGPML RKNQMKSTPW IKAYERNNVL SGLFCGLRGK AQIGKGMWAM PDLMADMYSQ KGDQLRAGAN TAWVPSPTAA TLHALHYHQT NVQSVQANIA QTEFNAEFEP LLDDLLTIPV AENANWSAQE IQQELDNNVQ GILGYVVRWV EQGIGCSKVP DIHNVALMED RATLRISSQH IANWLRHGIL TKEQVQASLE NMAKVVDQQN AGDPAYRPMA GNFANSCAFK AASDLIFLGV KQPNGYTEPL LHAWRLREKE SH |