missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant E. Coli MSRB Protein

A cDNA sequence encoding the MSRB was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12743-5ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15961399

  • 131.00 € / 5µg
Estimated Shipment: 15-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

MSRB Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MANKPSAEEL KKNLSEMQFY VTQNHGTEPP FTGRLLHNKR DGVYHCLICD APLFHSQTKY DSGCGWPSFY EPVSEESIRY IKDLSHGMQR IEIRCGNCDA HLGHVFPDGP QPTGERYCVN SASLRFTDGE NGEEING
Greater than 95.0% as determined by SDS-PAGE.
5 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
E. coli
His
MSRB protein solution (1 mg/ml) 20mM Tris-HCl buffer (pH 8.0), 20% glycerol 0.1M NaCl and 1mM DTT.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers