missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli Outer membrane protein 1
A DNA sequence encoding the Escherichia coli O157:H7 Outer membrane protein A, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7322-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_309068.1 | |
Outer membrane protein 1 | |
Research Use Only | |
ompA | |
Recombinant Protein | |
E. coli | |
WTNNIGDAHT IGTRPDNGML SLGVSYRFGQ GEAAPVVAPA PAPAPEVQTK HFTLKSDVLF NFNKATLKPE GQAALDQLYS QLSNLDPKDG SVVVLGYTDR IGSDAYNQGL SERRAQSVVD YLISKGIPAD KISARGMGES NPVTGNTCDN VKQRAALIDC LAPDRRVEIE VKGIKDVVTQ PQA | |
Greater than 90% as determined by SDS-PAGE. |
917123 | |
200 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
E. coli | |
His-SUMO | |
Tris-based buffer, 50% glycerol |