missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli Outer membrane protein 1
A DNA sequence encoding the Escherichia coli O157:H7 Outer membrane protein A, was expressed in the hosts and tags indicated.
309.00 € - 3260.00 €
Specifications
Accession Number | NP_309068.1 |
---|---|
Gene ID (Entrez) | 917123 |
Name | Outer membrane protein 1 |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16048190
|
enQuireBio™
QP7322-EC-100UG |
100 μg |
940.00 €
100µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16068190
|
enQuireBio™
QP7322-EC-1MG |
1 mg |
3260.00 €
1mg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16058190
|
enQuireBio™
QP7322-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16078190
|
enQuireBio™
QP7322-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16088190
|
enQuireBio™
QP7322-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16098190
|
enQuireBio™
QP7322-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_309068.1 | |
Outer membrane protein 1 | |
Not determined. | |
Partial (see sequence information for more details). | |
E. coli | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
917123 | |
Research Use Only | |
ompA | |
Recombinant Protein | |
E. coli | |
WTNNIGDAHT IGTRPDNGML SLGVSYRFGQ GEAAPVVAPA PAPAPEVQTK HFTLKSDVLF NFNKATLKPE GQAALDQLYS QLSNLDPKDG SVVVLGYTDR IGSDAYNQGL SERRAQSVVD YLISKGIPAD KISARGMGES NPVTGNTCDN VKQRAALIDC LAPDRRVEIE VKGIKDVVTQ PQA | |
Greater than 90% as determined by SDS-PAGE. |