missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant E. Coli PPA Protein

Product Code. p-7150995
Click to view available options
Quantity:
1 mg
20 μg
5 μg
Unit Size:
1mg
20µg
5µg
This item is not returnable. View return policy

Product Code. 15992539

Brand: enQuireBio™ QP131125ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A cDNA sequence encoding the PPA was constructed and used to recombinantly synthesize the protein.

Specifications

Name PPA Protein
Quantity 5 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Product Type Recombinant Protein
Cross Reactivity E. coli
Species E. coli
Protein Tag His
Sequence MGSSHHHHHH SSGLVPRGSH MSLLNVPAGK DLPEDIYVVI EIPANADPIK YEIDKESGAL FVDRFMSTAM FYPCNYGYIN HTLSLDGDPV DVLVPTPYPL QPGSVIRCRP VGVLKMTDEA GEDAKLVAVP HSKLSKEYDH IKDVNDLPEL LKAQIAHFFE HYKDLEKGKW VKVEGWENAE AAKAEIVASF ERAKNK
Buffer PPA protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 1mM DTT, 10% glycerol and 50mM NaCl.
Purity or Quality Grade Greater than 95.0% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.