missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli Probable diguanylate cyclase YedQ Protein
A DNA sequence encoding the Escherichia coli (strain K12) Probable diguanylate cyclase YedQ, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8608-ec-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_416465.2 | |
Probable diguanylate cyclase YedQ Protein | |
Research Use Only | |
yedQ | |
Recombinant Protein | |
E. coli | |
RRMVSNMYVL QSSLQWQAWH DTLTRLYNRG ALFEKARPLA KLCQTHQHPF SVIQVDLDHF KAINDRFGHQ AGDRVLSHAA GLISSSLRAQ DVAGRVGGEE FCVILPGASL TEAAEVAERI RLKLNEKEML IAKSTTIRIS ASLGVSSSEE TGDYDFEQLQ SLADRRLYLA KQAGRNRVFA SDNA | |
Greater than 90% as determined by SDS-PAGE. |
946471 | |
100 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
E. coli | |
GST | |
Tris-based buffer, 50% glycerol |