missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli Transketolase 1 Protein
A DNA sequence encoding the Escherichia coli (strain K12) Transketolase 1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7207-ec-100ug
Additional Details : Weight : 0.01000kg
Specifications
WP_000098614.1 | |
Transketolase 1 Protein | |
Research Use Only | |
tktA | |
Recombinant Protein | |
E. coli | |
MSSRKELANA IRALSMDAVQ KAKSGHPGAP MGMADIAEVL WRDFLKHNPQ NPSWADRDRF VLSNGHGSML IYSLLHLTGY DLPMEELKNF RQLHSKTPGH PEVGYTAGVE TTTGPLGQGI ANAVGMAIAE KTLAAQFNRP GHDIVDHYTY AFMGDGCMME GISHEVCSLA GTLKLGKLIA FYDDNGISID GHVEGWFTDD TAMRFEAYGW HVIRDIDGHD AASIKRAVEE ARAVTDKPSL LMCKTIIGFG SPNKAGTHDS HGAPLGDAEI ALTREQLGWK YAPFEIPSEI YAQWDAKEAG QAKESAWNEK FAAYAKAYPQ EAAEFTRRMK GEMPSDFDAK AKEFIAKLQA NPAKIASRKA SQNAIEAFGP LLPEFLGGSA DLAPSNLTLW SGSKAINEDA AGNYIHYGVR EFGMTAIANG ISLHGGFLPY TSTFLMFVEY ARNAVRMAAL MKQRQVMVYT HDSIGLGEDG PTHQPVEQVA SLRVTPNMST WRPCDQVESA VAWKYGVERQ DGPTALILSR QNLAQQERTE EQLANIARGG YVLKDCAGQP ELIFIATGSE VELAVAAYEK LTAEGVKARV VSMPSTDAFD KQDAAYRESV LPKAVTARVA VEAGIADYWY KYVGLNGAIV GMTTFGESAP AELLFEEFGF TVDNVVAKAK ELL | |
Greater than 90% as determined by SDS-PAGE. |
947420 | |
100 μg | |
Not determined. | |
Full Length | |
E. coli | |
His-SUMO | |
Tris-based buffer, 50% glycerol |