missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant EBV EA Protein

A cDNA sequence encoding the EBV EA was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11730-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15958168

  • 228.00 € / 100µg
Estimated Shipment: 27-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

EA Protein
Research Use Only
Recombinant Protein
E. coli
50mM Tris-HCl, 60mM NaCl, 50% glycerol and 10mM glutathione.
100 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
EBV
GST
EBV-Ea protein is >95% pure as determined by SDS-PAGE (coomassie staining).
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers