missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant EBV EA Protein
A cDNA sequence encoding the EBV EA was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11730-1mg
Additional Details : Weight : 0.01000kg
Specifications
EA Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
50mM Tris-HCl, 60mM NaCl, 50% glycerol and 10mM glutathione. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
EBV | |
GST | |
EBV-Ea protein is >95% pure as determined by SDS-PAGE (coomassie staining). |