missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant EBV p23 Protein
A cDNA sequence encoding the EBV p23 was constructed and used to recombinantly synthesize the protein.
373.00 € - 2175.00 €
Specifications
Name | p23 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | EBV |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15908188
|
enQuireBio™
QP11735-100UG |
100 μg |
373.00 €
100µg |
Estimated Shipment: 26-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15928188
|
enQuireBio™
QP11735-500UG |
500 μg |
1160.00 €
500µg |
Estimated Shipment: 26-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15918188
|
enQuireBio™
QP11735-1MG |
1 mg |
2175.00 €
1mg |
Estimated Shipment: 26-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
p23 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
EBV | |
Untagged | |
EBV- p23 protein is >95% pure as determined by 10% PAGE (coomassie staining). |
Research Use Only | |
Recombinant Protein | |
Epstein-Barr virus (EBV) | |
25mM glycine pH 9.6 and 50% glycerol. |