missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Epstein-Barr Apoptosis regulator BHRF1 Protein
A DNA sequence encoding the Epstein-Barr virus (strain B95-8) (HHV-4) (Human herpesvirus 4) Apoptosis regulator BHRF1, was expressed in the hosts and tags indicated.
421.00 € - 3590.00 €
Specifications
Accession Number | YP_401646.1 |
---|---|
Gene ID (Entrez) | 3783706 |
Name | Apoptosis regulator BHRF1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16097000
|
enQuireBio™
QP7005-10UG |
10 μg |
421.00 €
10µg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16037010
|
enQuireBio™
QP7005-50UG |
50 μg |
605.00 €
50µg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16087000
|
enQuireBio™
QP7005-100UG |
100 μg |
1025.00 €
100µg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16017010
|
enQuireBio™
QP7005-200UG |
200 μg |
1690.00 €
200µg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16027010
|
enQuireBio™
QP7005-500UG |
500 μg |
2260.00 €
500µg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16007010
|
enQuireBio™
QP7005-1MG |
1 mg |
3590.00 €
1mg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
YP_401646.1 | |
Apoptosis regulator BHRF1 Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Epstein-Barr | |
His | |
Tris-based buffer, 50% glycerol |
3783706 | |
Research Use Only | |
BHRF1 | |
Recombinant Protein | |
E. coli | |
MAYSTREILL ALCIRDSRVH GNGTLHPVLE LAARETPLRL SPEDTVVLRY HVLLEEIIER NSETFTETWN RFITHTEHVD LDFNSVFLEI FHRGDPSLGR ALAWMAWCMH ACRTLCCNQS TPYYVVDLSV RGMLEASEGL DG | |
Greater than 80% as determined by SDS-PAGE. |