missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Epstein-Barr DNA polymerase catalytic subunit Protein

Product Code. 16037960
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16037960 100 μg 100µg
16047960 10 μg 10µg
16087960 50 μg 50µg
16067960 200 μg 200µg
16077960 500 μg 500µg
16057960 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 16037960 Supplier enQuireBio™ Supplier No. QP7276ec100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A DNA sequence encoding the Epstein-Barr virus (strain B95-8) (HHV-4) (Human herpesvirus 4) DNA polymerase catalytic subunit, was expressed in the hosts and tags indicated.

Specifications

Accession Number YP_401712.1
Gene ID (Entrez) 3783681
Name DNA polymerase catalytic subunit Protein
Quantity 100 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol BALF5
Protein Length Partial (see sequence information for more details).
Product Type Recombinant Protein
Cross Reactivity Epstein-Barr
Species E. coli
Protein Tag His-SUMO
Sequence MSGGLFYNPF LRPNKGLLKK PDKEYLRLIP KCFQTPGAAG VVDVRGPQPP LCFYQDSLTV VGGDEDGKGM WWRQRAQEGT ARPEADTHGS PLDFHVYDIL ETVYTHEKCA VIPSDKQGYV VPCGIVIKLL GRRKADGASV CVNVFGQQAY FYASAPQGLD VEFAVLSALK ASTFDRRTPC RVSVEKVTRR SIMGYGNHAG DYHKITLSHP
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 90% as determined by SDS-PAGE.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.