missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Epstein-Barr Envelope glycoprotein 1

Product Code. p-7153986
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16067970 10 μg 10µg
16007980 50 μg 50µg
16057970 100 μg 100µg
16087970 200 μg 200µg
16097970 500 μg 500µg
16077970 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 16067970 Supplier enQuireBio™ Supplier No. QP7278ec10ug

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

A DNA sequence encoding the Epstein-Barr virus (strain B95-8) (HHV-4) (Human herpesvirus 4) Envelope glycoprotein L, was expressed in the hosts and tags indicated.

Specifications

Accession Number YP_401678.1
Gene ID (Entrez) 3783710
Name Envelope glycoprotein 1
Quantity 10 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol gL
Protein Length Full Length
Product Type Recombinant Protein
Cross Reactivity Epstein-Barr
Species E. coli
Protein Tag His-SUMO
Sequence NWAYPCCHVT QLRAQHLLAL ENISDIYLVS NQTCDGFSLA SLNSPKNGSN QLVISRCANG LNVVSFFISI LKRSSSALTG HLRELLTTLE TLYGSFSVED LFGANLNRYA WHRGG
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 90% as determined by SDS-PAGE.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.