missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Epstein-Barr Latent membrane protein 1
A DNA sequence encoding the Epstein-Barr virus (strain B95-8) (HHV-4) (Human herpesvirus 4) Latent membrane protein 2, was expressed in the hosts and tags indicated.
352.00 € - 3175.00 €
Specifications
Regulatory Status | Research Use Only |
---|---|
Endotoxin Concentration | Not determined. |
Product Type | Recombinant Protein |
Species | Yeast |
Protein Tag | His |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16037230
|
enQuireBio™
QP7054-YE-100UG |
100 μg |
1050.00 €
100µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16077230
|
enQuireBio™
QP7054-YE-500UG |
500 μg |
2535.00 €
500µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16047230
|
enQuireBio™
QP7054-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16067230
|
enQuireBio™
QP7054-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16087230
|
enQuireBio™
QP7054-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16057230
|
enQuireBio™
QP7054-YE-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
Research Use Only | |
Recombinant Protein | |
His |
Not determined. | |
Yeast | |
Tris-based buffer, 50% glycerol |